Learn More
Invitrogen™ Tex10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 720257
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Horse, Dog and Cat.
Testis-expressed sequence 10 protein (Tex10) functions as a component of the Five Friends of Methylated CHTOP (5FMC) complex. This complex is recruited to ZNF148 by methylated CHTOP leading to desumoylation of ZNF148 and subsequent transactivation of ZNF148 target genes. Tex10 is also involved in chromatin binding, maturation of LSU-rRNA, regulation of DNA-dependent DNA replication initiation, and rRNA processing.
Specifications
| Tex10 | |
| Polyclonal | |
| Unconjugated | |
| TEX10 | |
| 2610206N19Rik; 2810462N03Rik; bA208F1.2; BC006867; clone 18330; Ipi1; L18; Nbla10363; testis expressed 10; testis expressed gene 10; Testis-expressed protein 10; testis-expressed sequence 10 protein; TEX10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 269536, 298065, 54881 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
| Q3URQ0, Q9NXF1 | |
| TEX10 | |
| Peptides corresponding to Human TEX10 (aa 240-258, 13-25). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.