missing translation for 'onlineSavingsMsg'
Learn More

Tetronic Acid, TRC

Product Code. 30456305
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456305 100 mg 100mg
30454465 50 mg 50mg
30449952 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456305 Supplier Toronto Research Chemicals Supplier No. TRCT309515100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4971-56-6
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
Synonym Furan-2,4-dione
IUPAC Name oxolane-2,4-dione
SMILES O=C1COC(=O)C1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C4H4O3/c5-3-1-4(6)7-2-3/h1-2H2
Formula Weight 100.016
Chemical Name or Material Tetronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.