missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetrazolium Violet, 98+%

Product Code. 11478616
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478616 1 g 1g
11488616 5 g 5g
11498616 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478616 Supplier Thermo Scientific Chemicals Supplier No. A16008.03
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
This item is not returnable. View return policy

CAS: 1719-71-7 | C23H17ClN4 | 384.867 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1719-71-7
Molecular Formula C23H17ClN4
Molecular Weight (g/mol) 384.867
MDL Number MFCD00011875
InChI Key RONADMZTCCPLEF-UHFFFAOYSA-M
Synonym tetrazolium violet, violet tetrazolium, tetrazolium purple, tv, 2,5-diphenyl-3 1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-alpha-naphthyl-2h-tetrazolium chloride, 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium chloride, 2h-tetrazolium, 3-1-naphthyl-2,5-diphenyl-, chloride, tetrazolium violet at
PubChem CID 74395
ChEBI CHEBI:75193
IUPAC Name 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium;chloride
SMILES C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC4=CC=CC=C43)C5=CC=CC=C5.[Cl-]

Specifications

Melting Point 247°C to 249°C
Quantity 1 g
Formula Weight 384.87
Percent Purity ≥98%
Chemical Name or Material Tetrazolium Violet

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.