missing translation for 'onlineSavingsMsg'
Learn More

Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate, 95%

Product Code. 11327308
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327308 250 mg 250mg
11317308 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327308 Supplier Thermo Scientific Alfa Aesar Supplier No. 030538.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122451-08-5 | C44H54N4Na4O24S4 | 1243.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122451-08-5
Molecular Formula C44H54N4Na4O24S4
Molecular Weight (g/mol) 1243.119
MDL Number MFCD00151011
InChI Key CDOFYZDFUGMKDJ-UHFFFAOYSA-N
Synonym 4,4',4,4'-porphine-5,10,15,20-tetrayl tetrakis benzenesulfonic acid tetrasodium salt hydrate
PubChem CID 131869494
IUPAC Name sodium;4-[10,15,20-tris(4-sulfophenyl)-21,24-dihydroporphyrin-5-yl]benzenesulfonic acid;dodecahydrate
SMILES C1=CC(=CC=C1C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)S(=O)(=O)O)C8=CC=C(C=C8)S(=O)(=O)O)C9=CC=C(C=C9)S(=O)(=O)O)N3)S(=O)(=O)O.O.O.O.O.O.O.O.O.O.O.O.O.[Na].[Na].[Na].[Na]

Specifications

Quantity 250 mg
Beilstein 5488862
Formula Weight 1239.11 (1022.87 Anhydrous)
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.