missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium hydroxide, 25% in water

Product Code. 10289460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10289460 100 g Plastic bottle 100g
10576792 25 g Plastic bottle 25g
11975451 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10289460 Supplier Thermo Scientific Acros Supplier No. 420731000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4499-86-9 | C12H29NO | 203.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4499-86-9 , 7732-18-5
Molecular Formula C12H29NO
Molecular Weight (g/mol) 203.37
MDL Number MFCD00009360
InChI Key LPSKDVINWQNWFE-UHFFFAOYSA-M
Synonym tetrapropylammonium hydroxide, tetra-n-propylammonium hydroxide, 1-propanaminium, n,n,n-tripropyl-, hydroxide, tpaoh, tetrapropylammonium oxide, unii-t686luy7nk, tetrapropylazanium hydroxide, t686luy7nk, ammonium, tetrapropyl-, hydroxide, n,n,n-tripropyl-1-propanaminium hydroxide
PubChem CID 20586
SMILES [OH-].CCC[N+](CCC)(CCC)CCC

Specifications

Boiling Point 100.0°C to 102.0°C
Linear Formula (CH3CH2CH2)4NOH
Density 1.0000g/mL
Solubility Information Solubility in water: soluble in water.
IUPAC Name tetrapropylazanium;hydroxide
Formula Weight 203.37
Packaging Plastic bottle
Specific Gravity 1
Quantity 100 g
Chemical Name or Material Tetrapropylammonium hydroxide, 25% in Water

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.