missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium chloride, 94%

Product Code. p-3779678 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10483931 25 g Plastic bottle 25g
10147143 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10483931 Supplier Thermo Scientific Acros Supplier No. 420710250
Add to basket
Edge
This item is not returnable. View return policy

CAS: 5810-42-4 | C12H28ClN | 221.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5810-42-4
Molecular Formula C12H28ClN
Molecular Weight (g/mol) 221.81
MDL Number MFCD00038729
InChI Key FBEVECUEMUUFKM-UHFFFAOYSA-M
Synonym tetrapropylammonium chloride, tetrapropyl ammonium chloride, tetra-n-propylammonium chloride, 1-propanaminium, n,n,n-tripropyl-, chloride, 1-propanaminium, n,n,n-tripropyl-, chloride 1:1, n,n,n-tripropylpropan-1-aminium chloride, tetrapropylazanium chloride, acmc-209m2k, ksc491c1h, tetra-n-propylammonium chloride 1g
PubChem CID 79880
IUPAC Name tetrapropylazanium;chloride
SMILES CCC[N+](CCC)(CCC)CCC.[Cl-]

Specifications

Melting Point 240.0°C to 242.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 92% min. (Argentometry)
Packaging Plastic bottle
Linear Formula (CH3CH2CH2)4NCl
Quantity 25 g
Formula Weight 221.81
Percent Purity 94%
Physical Form Crystals
Chemical Name or Material Tetrapropylammonium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.