missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylphosphonium iodide, 98+%, Thermo Scientific Chemicals

Product Code. 11494553
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2065-67-0
Molecular Formula C24H20IP
Molecular Weight (g/mol) 466.30
MDL Number MFCD00011917
InChI Key AEFPPQGZJFTXDR-UHFFFAOYSA-M
Synonym tetraphenylphosphonium iodide, tetraphenylphosphoniumiodide, tetraphenylphosphanium iodide, acmc-1cpca
PubChem CID 2724164
IUPAC Name tetraphenylphosphanium;iodide
SMILES [I-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 345°C to 350°C
Quantity 5 g
Beilstein 3922216
Sensitivity Light sensitive
Formula Weight 466.31
Percent Purity ≥98%
Chemical Name or Material Tetraphenylphosphonium iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'