missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylphosphonium iodide, 98+%

Product Code. 11414563
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414563 100 g 100g
11494553 5 g 5g
11404563 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414563 Supplier Thermo Scientific Alfa Aesar Supplier No. A11539.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2065-67-0 | C24H20IP | 466.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2065-67-0
Molecular Formula C24H20IP
Molecular Weight (g/mol) 466.30
MDL Number MFCD00011917
InChI Key AEFPPQGZJFTXDR-UHFFFAOYSA-M
Synonym tetraphenylphosphonium iodide, tetraphenylphosphoniumiodide, tetraphenylphosphanium iodide, acmc-1cpca
PubChem CID 2724164
IUPAC Name tetraphenylphosphanium;iodide
SMILES [I-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 345°C to 350°C
Quantity 100 g
Beilstein 3922216
Sensitivity Light sensitive
Formula Weight 466.31
Percent Purity ≥98%
Chemical Name or Material Tetraphenylphosphonium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.