missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylphosphonium chloride, 98%

Product Code. 10502421 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10502421 50 g Glass bottle 50g
10388240 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10502421 Supplier Thermo Scientific Acros Supplier No. 204030500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2001-45-8 | C24H20ClP | 374.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2001-45-8
Molecular Formula C24H20ClP
Molecular Weight (g/mol) 374.85
MDL Number MFCD00011916
InChI Key WAGFXJQAIZNSEQ-UHFFFAOYSA-M
Synonym tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co
PubChem CID 164911
IUPAC Name tetraphenylphosphanium;chloride
SMILES [Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 275.0°C to 280.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C6H5)4PCl
Quantity 50 g
Beilstein 16, II, 373
Solubility Information Solubility in water: soluble.
Formula Weight 374.84
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenylphosphonium chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Tetraphenylphosphonium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.