missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylphosphine imide, 97%

Product Code. p-7043374
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11390064

Brand: Thermo Scientific Alfa Aesar L00496.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2325-27-1 | C24H20NP | 353.41 g/mol

Has been used as a protected form of aniline for the preparation of monoalkylanilines by alkylation and hydrolysis and as an intermediate for the preparation of isocyanates and isothiocyanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Has been used as a protected form of aniline for the preparation of monoalkylanilines by alkylation and hydrolysis and as an intermediate for the preparation of isocyanates and isothiocyanates.

Solubility
Insoluble in water. Soluble in chloroform 25 mg/mL.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2325-27-1
Molecular Formula C24H20NP
Molecular Weight (g/mol) 353.41
MDL Number MFCD00003044
InChI Key PTLOPIHJOPWUNN-UHFFFAOYSA-N
Synonym tetraphenylphosphine imide, n-triphenylphosphoranylidene aniline, phenylimino triphenylphosphorane, phosphine imide, tetraphenyl, benzenamine, n-triphenylphosphoranylidene, tetraphenylphosphinimine, triphenylphosphine phenylimide, benzenamine,n-triphenylphosphoranylidene, n,1,1,1-tetraphenyl-??-phosphanimine, triphenyl phenylimino
PubChem CID 75352
IUPAC Name triphenyl(phenylimino)-$l^{5}-phosphane
SMILES C1=CC=C(C=C1)N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 128°C to 132°C
Odor Odorless
Linear Formula (C6H5)3P=N(C6H5)
Quantity 1 g
Beilstein 753032
Sensitivity Moisture sensitive
Solubility Information Insoluble in water. Soluble in chloroform 25mg/mL.
Formula Weight 353.4
Percent Purity 97%
Chemical Name or Material Tetraphenylphosphine imide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.