missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylgermanium, 98%, Thermo Scientific™

Product Code. 11378737
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378737 2 g 2g
11368737 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378737 Supplier Thermo Scientific Alfa Aesar Supplier No. 035842.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1048-05-1
Molecular Formula C24H20Ge
Molecular Weight (g/mol) 381.054
MDL Number MFCD00002997
InChI Key ILEXMONMGUVLRM-UHFFFAOYSA-N
Synonym tetraphenylgermanium, germane, tetraphenyl, tetraphenylgermane, germanium tetraphenyl
PubChem CID 66103
IUPAC Name tetraphenylgermane
SMILES C1=CC=C(C=C1)[Ge](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 231°C to 233°C
Color White
Quantity 2 g
Formula Weight 381.03
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Tetraphenylgermanium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.