missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylcyclopentadienone, 98%

Product Code. 11455083
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455083 100 g 100g
11435083 5 g 5g
11445083 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455083 Supplier Thermo Scientific Alfa Aesar Supplier No. A12756.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 479-33-4 | C29H20O | 384.478 g/mol

It is an easily made building block for many organic and organometallic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an easily made building block for many organic and organometallic compounds.

Solubility
Soluble in Benzene. Insoluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-33-4
Molecular Formula C29H20O
Molecular Weight (g/mol) 384.478
MDL Number MFCD00001407
InChI Key PLGPSDNOLCVGSS-UHFFFAOYSA-N
Synonym tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone
PubChem CID 68068
IUPAC Name 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one
SMILES C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Melting Point 217°C to 219°C
Quantity 100 g
Beilstein 683115
Solubility Information Soluble in Benzene. Insoluble in water.
Formula Weight 384.48
Percent Purity 98%
Chemical Name or Material Tetraphenylcyclopentadienone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.