missing translation for 'onlineSavingsMsg'
Learn More

Tetrapentylammonium bromide, 99+%

Product Code. 10116733 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10116733 100 g Glass bottle 100g
10441071 5 g Glass bottle 5g
10246800 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10116733 Supplier Thermo Scientific Acros Supplier No. 221481000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 866-97-7 | C20H44BrN | 378.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866-97-7
Molecular Formula C20H44BrN
Molecular Weight (g/mol) 378.48
MDL Number MFCD00011856
InChI Key SPALIFXDWQTXKS-UHFFFAOYSA-M
Synonym tetrapentylammonium bromide, tetraamylammonium bromide, tetra-n-amylammonium bromide, 1-pentanaminium, n,n,n-tripentyl-, bromide 1:1, tetrapentylazanium bromide, n,n,n-tripentylpentan-1-aminium bromide, tetraamylammoniumbromide, 1-pentanaminium, n,n,n-tripentyl-, bromide, acmc-209qb0, tetrapentyl ammonium bromide
PubChem CID 70086
SMILES [Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC

Specifications

Melting Point 100°C to 102°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula [CH3(CH2)4]4NBr
Quantity 100 g
Beilstein 04, III, 331
IUPAC Name tetrapentylazanium;bromide
Formula Weight 378.47
Percent Purity 99+%
Physical Form Shiny Flakes
Chemical Name or Material Tetrapentylammonium bromide
Show More Show Less
compliance-icons
Product Identifier
  • Tetrapentylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.