missing translation for 'onlineSavingsMsg'
Learn More

Tetraoctyltin (>85%), TRC

Product Code. 30005825
Change view
Click to view available options
Quantity:
5 mL
25 mL
50 mL
Unit Size:
25mL
50mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005825 50 mL 50mL
30006574 25 mL 25mL
17931345 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005825 Supplier Toronto Research Chemicals Supplier No. TRCT31045350ML
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3590-84-9
Molecular Formula C32 H68 Sn
Molecular Weight (g/mol) 571.59
Synonym Tin, tetraoctyl- (6CI,7CI), Tetraoctylstannane, NSC 65527, Tetra-n-octylstannane, Tetra-n-octyltin, Tetraoctyltin
IUPAC Name tetraoctylstannane
SMILES CCCCCCCC[Sn](CCCCCCCC)(CCCCCCCC)CCCCCCCC

Specifications

Quantity 50 mL
InChI Formula InChI=1S/4C8H17.Sn/c4*1-3-5-7-8-6-4-2;/h4*1,3-8H2,2H3;
Formula Weight 572.4343
Percent Purity >95%
Chemical Name or Material Tetraoctyltin
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.