missing translation for 'onlineSavingsMsg'
Learn More

Tetraoctyltin 1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30322903
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322903 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322903 Supplier Dr. Ehrenstorfer Supplier No. DREV17414750DI1000
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetraoctyltin
CAS 3590-84-9
Analyte or Component Names Tetraoctyltin
Shipping Condition +20°C
Molecular Formula C32 H68 Sn
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Tetraoctylstannane, NSC 65527, Tetra-n-octylstannane, Tetra-n-octyltin, Tin, tetraoctyl- (6CI,7CI)
InChI Formula InChI=1S/4C8H17.Sn/c4*1-3-5-7-8-6-4-2;/h4*1,3-8H2,2H3;
SMILES CCCCCCCC[Sn](CCCCCCCC)(CCCCCCCC)CCCCCCCC
IUPAC Name tetraoctylstannane
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 571.59
Quantity 5 mL
Formula Weight 572.4343 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.