missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetranitro blue tetrazolium chloride, 97%

Product Code. 15453739
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 15453739

Brand: Thermo Scientific Chemicals J61080.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Redox indicator for enzymes | CAS: 1184-43-6 | C40H28Cl2N12O10 | 907.64 g/mol

Redox indicator for enzymes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-43-6
Molecular Formula C40H28Cl2N12O10
Molecular Weight (g/mol) 907.64
MDL Number MFCD00036338
InChI Key VCESGVLABVSDRO-UHFFFAOYSA-L
Synonym TNBT
PubChem CID 129893520
SMILES [Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Beilstein 3897475
IUPAC Name 2-{4'-[3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl}-3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Formula Weight 999.78
Percent Purity 97%
Color Yellow
Quantity 100 mg
Physical Form Powder
Chemical Name or Material Tetranitro blue tetrazolium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.