missing translation for 'onlineSavingsMsg'
Learn More

Tetrandrine, MedChemExpress

Product Code. 16479855
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479855 250 mg 250mg
16456316 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479855 Supplier Medchem Express Supplier No. HY13764

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tetrandrine (NSC-77037; d-Tetrandrine) is a bis-benzyl-isoquinoline alkaloid, which inhibits voltage-gated Ca2+ current (ICa) and Ca2+-activated K+ current.

Tetrandrine (NSC-77037

Specifications

Chemical Name or Material Tetrandrine
CAS 518-34-3
Color White
Purity Grade Notes Research
Molecular Formula C38H42N2O6
Quantity 250 mg
Synonym NSC-77037 d-Tetrandrine
Solubility Information DMSO : 5 mg/mL (8.03 mM; Need ultrasonic and warming)
SMILES COC1=CC(CCN(C)[C@@]2([H])CC3=CC(O4)=C(OC)C=C3)=C2C(OC5=C(OC)C=C6C([C@]([H])(CC7=CC=C4C=C7)N(C)CC6)=C5)=C1OC
Molecular Weight (g/mol) 622.75
Formula Weight 622.75
Percent Purity 99.49%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.