missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TETRAN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116158
This item is not returnable.
View return policy
Description
TETRAN Polyclonal Antibody for Western Blot
TETRAN's overexpression in cultured human cells caused resistance to some NSAIDs (Non-steroidal anti-inflammatory drugs), suggesting that TETRAN is an efflux pump for some NSAIDs.
Specifications
| TETRAN | |
| Polyclonal | |
| Unconjugated | |
| MFSD10 | |
| 0610009O03Rik; major facilitator superfamily domain containing 10; major facilitator superfamily domain-containing protein 10; MFSD10; RGD1311900; Tetracycline transporter-like protein; TETRAN; TETTRAN | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 10227 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q14728 | |
| MFSD10 | |
| KLH conjugated synthetic peptide between 245-274 amino acids from the Central region of human MFS10. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction