missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyltin, 98%

Product Code. 10265590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265590 50 g Glass bottle 50g
10001950 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10265590 Supplier Thermo Scientific Acros Supplier No. 163980500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 594-27-4 | C4H12Sn | 178.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-27-4
Molecular Formula C4H12Sn
Molecular Weight (g/mol) 178.83
MDL Number MFCD00008278
InChI Key VXKWYPOMXBVZSJ-UHFFFAOYSA-N
Synonym tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
PubChem CID 11661
ChEBI CHEBI:30420
IUPAC Name tetramethylstannane
SMILES C[Sn](C)(C)C

Specifications

Melting Point -54.0°C
Color Colorless
Density 1.2900g/mL
Boiling Point 74.0°C to 75.0°C
Flash Point −12°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)4Sn
Refractive Index 1.4400 to 1.4420
Quantity 50 g
Beilstein 04, 631
Specific Gravity 1.29
Solubility Information Solubility in water: immiscible.
Formula Weight 178.83
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetramethyltin, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethyltin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.