missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyltin, 98%

Product Code. 11334839
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334839 50 g 50g
11354839 2 g 2g
11344839 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334839 Supplier Thermo Scientific Alfa Aesar Supplier No. 071145.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 594-27-4 | C4H12Sn | 178.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-27-4
Molecular Formula C4H12Sn
Molecular Weight (g/mol) 178.85
MDL Number MFCD00008278
InChI Key VXKWYPOMXBVZSJ-UHFFFAOYSA-N
Synonym tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
PubChem CID 11661
ChEBI CHEBI:30420
IUPAC Name tetramethylstannane
SMILES C[Sn](C)(C)C

Specifications

Melting Point -54°C
Density 1.291 g/mL
Boiling Point 74°C to 75°C
Flash Point −12°C (9°F)
Odor Unpleasant
Refractive Index 1.441
Quantity 50 g
UN Number UN3384
Solubility Information Insoluble in water; Soluble in organic solvents
Formula Weight 178.83
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetramethyltin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.