missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Tetramethylrhodamine (TAMRA) Alkyne (5-Carboxytetramethylrhodamine, Propargylamide), 5-isomer
Description
The red-fluorescent tetramethylrhodamine (TAMRA) alkyne can be reacted with azides via a copper-catalyzed click reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.
Specifications
Specifications
| Format | Solid |
| Quantity | 0.5 mg |
| Product Type | Tetramethylrhodamine Alkyne |
| Molecular Weight (g/mol) | 467.53 Da |
| Solubility | DMSO (Dimethylsulfoxide) |
| Content And Storage | Store at ≤-20°C, desiccated and protected from light. |
| Chemical Reactivity | Azide |
| Detection Method | Fluorescence |
| Reactive Group | Alkyne |
| Shipping Condition | Room Temperature |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?