missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylphosphonium Bromide, TRC

Product Code. 30456454
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456454 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456454 Supplier Toronto Research Chemicals Supplier No. TRCT890280100MG
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetramethylphosphonium Bromide
CAS 4519-28-2
Molecular Formula C4 H12 P . Br
Quantity 100 mg
Synonym Phosphonium','tetramethyl-','bromide (8CI','9CI), Tetramethylphosphonium bromide (7CI), NSC 617067
InChI Formula InChI=1S/C4H12P.BrH/c1-5(2,3)4;/h1-4H3;1H/q+1;/p-1
SMILES [Br-].C[P+](C)(C)C
IUPAC Name tetramethylphosphanium;bromide
Molecular Weight (g/mol) 171.016
Formula Weight 169.986
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.