missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylphosphonium bromide, 97%

Product Code. 11419026
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419026 5 g 5g
11409026 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419026 Supplier Thermo Scientific Alfa Aesar Supplier No. A17907.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4519-28-2 | C4H12BrP | 171.02 g/mol

It is used as a pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate and in chemical research.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4519-28-2
Molecular Formula C4H12BrP
Molecular Weight (g/mol) 171.02
MDL Number MFCD00011802
InChI Key ZTXFOCMYRCGSMU-UHFFFAOYSA-M
Synonym tetramethylphosphonium bromide, tetramethylphosphanium bromide, acmc-1ai8x, ch3 4pbr, tetramethyl phosphonium bromide, phosphonium, tetramethyl-, bromide
PubChem CID 357594
IUPAC Name tetramethylphosphanium;bromide
SMILES [Br-].C[P+](C)(C)C

Specifications

Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 171.03
Percent Purity 98%
Chemical Name or Material Tetramethylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.