missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyldisilazane, 97%

Product Code. 11476837
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476837 50 g 50g
11466837 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476837 Supplier Thermo Scientific Alfa Aesar Supplier No. A14304.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15933-59-2 | C4H13NSi2 | 131.325 g/mol

Tetramethyldisilazane is used as a gas chromatographic derivatizing reagent. Further, it reacts with phenol to prepare dimethylphenoxysilane. In addition, it is used in electronic, polymer and pharmaceutical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyldisilazane is used as a gas chromatographic derivatizing reagent. Further, it reacts with phenol to prepare dimethylphenoxysilane. In addition, it is used in electronic, polymer and pharmaceutical industries.

Solubility
Miscible with common organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15933-59-2
Molecular Formula C4H13NSi2
Molecular Weight (g/mol) 131.325
MDL Number MFCD00025626
InChI Key GJWAPAVRQYYSTK-UHFFFAOYSA-N
Synonym 1,1,3,3-tetramethyldisilazane, bis dimethylsilyl amine, tetramethyldisilazane, tetramethyl disilazane, tmds, silanamine, n-dimethylsilyl-1,1-dimethyl, disilazane, 1,1,3,3-tetramethyl, c4h15nsi2, n-dimethylsilyl-1,1-dimethylsilylamine, 1,3,3-tetramethyldisilazane
PubChem CID 6327317
IUPAC Name [(dimethyl-$l^{3}-silanyl)amino]-dimethylsilicon
SMILES C[Si](C)N[Si](C)C

Specifications

Boiling Point 99°C to 100°C
Quantity 50 g
UN Number UN2924
Beilstein 741869
Flash Point −3°C (26°F)
Solubility Information Miscible with common organic solvents.
Formula Weight 133.34
Percent Purity 97%
Odor Ammonia-like
Refractive Index 1.4055
Sensitivity Moisture sensitive
Density 0.768
Chemical Name or Material Tetramethyldisilazane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.