missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Triacetoxyborohydride (>90%), TRC

Product Code. 30424938
Change view
Click to view available options
Quantity:
25 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424938 25 g
30420136 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424938 Supplier Toronto Research Chemicals Supplier No. TRCT29872025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109704-53-2
Molecular Formula C10H22BNO6
Molecular Weight (g/mol) 263.1
Synonym N,N,N-Trimethylmethanaminium (T-4)-tris(Acetato-O)hydroborate(1-), (T-4)-tris(Acetato-O)hydroborate(1-) N,N,N-trimethylmethanaminium, (T-4)-tris(Acetato-kO)hydroborate(1-) N,N,N-trimethylmethanaminium, Me4NHB(OAc)3, Tetramethylammonium Triacetoxyborohydride
IUPAC Name tetramethylazanium;triacetyloxyboranuide
SMILES CC(=O)O[BH-](OC(=O)C)OC(=O)C.C[N+](C)(C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H10BO6.C4H12N/c1-4(8)11-7(12-5(2)9)13-6(3)10;1-5(2,3)4/h7H,1-3H3;1-4H3/q-1;+1
Formula Weight 263.154
Chemical Name or Material Tetramethylammonium Triacetoxyborohydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.