missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Tetrafluoroborate, TRC

Product Code. 30451837
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451837 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451837 Supplier Toronto Research Chemicals Supplier No. TRCT889868500MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Tetramethylammonium Tetrafluoroborate
CAS 661-36-9
Molecular Formula C4H12BF4N
Quantity 500 mg
Synonym Methanaminium','N,N,N-trimethyl-','tetrafluoroborate(1-) (1:1) (ACI), Ammonium compounds','substituted','tetramethyl-','tetrafluoroborate (7CI), Ammonium','tetramethyl-','tetrafluoroborate(1-) (8CI), Borate(1-)','tetrafluoro-','N,N,N-trimethylmethanaminium (ZCI), Borate(1-)','tetrafluoro-','tetramethylammonium (8CI), Methanaminium','N,N,N-trimethyl-','tetrafluoroborate(1-) (9CI), Tetramethylammonium fluoroborate (6CI), [Me4N][BF4], Tetramethylammonium tetrafluoroborate, Tetramethylammonium tetrafluoroborate(1-)
InChI Formula InChI=1S/C4H12N.BF4/c1-5(2,3)4;2-1(3,4)5/h1-4H3;/q+1;-1
SMILES C[N+](C)(C)C.F[B-](F)(F)F
IUPAC Name tetramethylazanium;tetrafluoroborate
Molecular Weight (g/mol) 160.95
Formula Weight 161.0999
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.