missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide Solution, 25% in H2O, TraceSELECT™, Honeywell™

Product Code. 15616710 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15616710 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15616710 Supplier Honeywell-Fluka Supplier No. 68556250ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75-59-2
Density 1.020g/mL at 20°C
Molecular Formula C4H13NO
Linear Formula (CH3)4N(OH)
MDL Number MFCD00008280
Quantity 250 mL
UN Number UN1835
Beilstein 3558708
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES C[N+](C)(C)C.[OH-]
IUPAC Name tetramethylazanium;hydroxide
Molecular Weight (g/mol) 91.154
PubChem CID 60966
Formula Weight 91.15g/mol
Chemical Name or Material Tetramethylammonium hydroxide solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.