missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide (25% w/w solution in Water), TRC

Product Code. 30424770
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424770 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424770 Supplier Toronto Research Chemicals Supplier No. TRCT300155250ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetramethylammonium Hydroxide
CAS 75-59-2
Molecular Formula C4 H12 N . H O
Quantity 250 mL
Synonym N,N,N-Trimethylmethanaminium Hydroxide, Methanaminium','N,N,N-trimethyl-','hydroxide (1:1) (ACI), Ammonium','tetramethyl-','hydroxide (8CI), Methanaminium','N,N,N-trimethyl-','hydroxide (9CI), Tetramethylammonium hydroxide (6CI), AR 300-49, AZ 300MIF, AZ 726, AZ 726MIF, AZ-MIF 300, CD 26, CPD 18, L 09658, LDD 26, LDD 26W, Megaposit CD 14, Megaposit MF 24A, MF 24A, MF 319, MF 321, MF-CD 26, Microposit CD 26, Microposit MF 319, Microposit MF 321, Microposit MF-CD 26, MIF 726, NMD 3, NMW-W, OCG 934, OCG 945, OCG-OPD 262, OPD 262, OPD 5262, Photoresist ma-D 525, PRS 2000, RD 6, RD 6 (developer), TMAH, TMAH 25, Tokuso SD 20, Tokuso SD 25
InChI Formula InChI=1S/C4H12N.H2O/c1-5(2,3)4;/h1-4H3;1H2/q+1;/p-1
SMILES [OH-].C[N+](C)(C)C
IUPAC Name tetramethylazanium;hydroxide
Molecular Weight (g/mol) 91.15
Formula Weight 91.0997
Concentration or Composition (by Analyte or Components) 0.25
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.