Learn More
Tetramethylammonium hydroxide, 25% w/w aq. soln., Electronic Grade, 99.9999% (metals basis), Thermo Scientific Chemicals
£49.60 - £141.00
Chemical Identifiers
CAS | 75-59-2 |
---|---|
Molecular Formula | C4H13NO |
Molecular Weight (g/mol) | 91.154 |
MDL Number | MFCD00008280 |
InChI Key | WGTYBPLFGIVFAS-UHFFFAOYSA-M |
Synonym | tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide |
PubChem CID | 60966 |
IUPAC Name | tetramethylazanium;hydroxide |
SMILES | C[N+](C)(C)C.[OH-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11347987
|
Thermo Scientific Alfa Aesar
020932.AK |
250 mL |
£49.60
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357987
|
Thermo Scientific Alfa Aesar
020932.K2 |
1 L |
£141.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetramethylammonium hydroxide is used to produce tetramethyl-ammonium azide. It is used as an anisotropic etchant of silicon, basic solvent in the development of acidic photoresist in photolithography process, as surfactant in the synthesis of ferrofluid and as a polarographic reagent. It finds application in the production of organic silicon, where it is used for computer silicon wafer surface brightener and cleaning agent. It is also involved in the purification of some metallic elements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-59-2 | |
91.154 | |
WGTYBPLFGIVFAS-UHFFFAOYSA-M | |
60966 | |
C[N+](C)(C)C.[OH-] |
C4H13NO | |
MFCD00008280 | |
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide | |
tetramethylazanium;hydroxide |
Specifications
75-59-2 | |
1.016 g/mL | |
C4H13NO | |
1.384 | |
UN1835 | |
Air Sensitive | |
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide | |
WGTYBPLFGIVFAS-UHFFFAOYSA-M | |
tetramethylazanium;hydroxide | |
60966 | |
99.9999% | |
25% w/w aq. soln. | |
(metals basis) |
Colorless to Yellow | |
∼102°C | |
(CH3)4NOH | |
MFCD00008280 | |
3558708 | |
14,9224 | |
Miscible with water. | |
C[N+](C)(C)C.[OH-] | |
91.154 | |
91.15 | |
Electronic | |
Liquid | |
Tetramethylammonium hydroxide |
Safety and Handling
GHS H Statement
H300-H310-H370-H372-H314
Fatal if swallowed.
Fatal in contact with skin.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H310-H314-H370-H372
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TETRAMETHYLAMMONIUM HYDROXIDE SOLUTION
missing translation for 'einecsNumber' : 200-882-9
missing translation for 'rtecsNumber' : PA0875000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Tetramethylammonium hydroxide, 25% w/w aqueous soluion, Electronic Grade
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H300 + H310-Fatal if swallowed or in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H370-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only