missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydrogensulfate, 99+%, HPLC grade
GREENER_CHOICE

Product Code. 10415125 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10415125 50 g Glass bottle 50g
10565291 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10415125 Supplier Thermo Scientific Supplier No. 343830500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 80526-82-5 | C4H13NO4S | 171.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80526-82-5
Molecular Formula C4H13NO4S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00036149
InChI Key DWTYPCUOWWOADE-UHFFFAOYSA-M
Synonym tetramethylammonium hydrogen sulfate, tetramethylammonium bisulfate, tetramethylammonium hydrogen sulphate, tetramethylammonium hydrogensulfate, hydrogen sulfate; tetramethylazanium, methanaminium, n,n,n-trimethyl-, sulfate 1:1, hydrogen sulfate; tetramethylammonium, acmc-209pjn, ch3 4n hso4 .h2o, tetramethylammonium bisulfate solution
PubChem CID 157340
IUPAC Name hydrogen sulfate;tetramethylazanium
SMILES C[N+](C)(C)C.OS(=O)(=O)[O-]

Specifications

Absorbance (0.005M in water), (0.005 M in water) at 0nm, 0.05 max. at 220nm, 0.15 max. at 200nm
Melting Point 295.0°C to 297.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 50 g
Formula Weight 171.21
Percent Purity 99+%
Grade HPLC
Physical Form Powder
Chemical Name or Material Tetramethylammonium hydrogensulfate
Show More Show Less
compliance-icons
Product Identifier
  • Tetramethylammonium hydrogensulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.