missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hexafluorophosphate, 99%

Product Code. 11448067
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448067 100 g 100g
11438067 25 g 25g
11458067 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448067 Supplier Thermo Scientific Alfa Aesar Supplier No. A17467.22
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 558-32-7 | C4H12F6NP | 219.111 g/mol

It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 558-32-7
Molecular Formula C4H12F6NP
Molecular Weight (g/mol) 219.111
MDL Number MFCD00012012
InChI Key ZFKULDQWLXAKIM-UHFFFAOYSA-N
Synonym tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci
PubChem CID 11209
IUPAC Name tetramethylazanium;hexafluorophosphate
SMILES C[N+](C)(C)C.F[P-](F)(F)(F)(F)F

Specifications

Melting Point >300°C
Odor Odorless
Linear Formula (CH3)4NPF6
Quantity 100 g
UN Number UN2923
Beilstein 3730394
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 219.1
Percent Purity ≥99%
Chemical Name or Material Tetramethylammonium hexafluorophosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.