missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hexafluorophosphate, 99%

Product Code. 11438067
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438067 25 g 25g
11448067 100 g 100g
11458067 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438067 Supplier Thermo Scientific Alfa Aesar Supplier No. A17467.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 558-32-7 | C4H12F6NP | 219.111 g/mol

It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 558-32-7
Molecular Formula C4H12F6NP
Molecular Weight (g/mol) 219.111
MDL Number MFCD00012012
InChI Key ZFKULDQWLXAKIM-UHFFFAOYSA-N
Synonym tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci
PubChem CID 11209
IUPAC Name tetramethylazanium;hexafluorophosphate
SMILES C[N+](C)(C)C.F[P-](F)(F)(F)(F)F

Specifications

Melting Point >300°C
Odor Odorless
Linear Formula (CH3)4NPF6
Quantity 25 g
UN Number UN2923
Beilstein 3730394
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 219.1
Percent Purity ≥99%
Chemical Name or Material Tetramethylammonium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • Tetramethylammonium hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tetramethylammoniumhexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.