missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium formate, 30% w/w aq. soln.

Product Code. 11346149
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346149 500 mL 500mL
11315477 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346149 Supplier Thermo Scientific Alfa Aesar Supplier No. 042710.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59138-84-0 | C5H13NO2 | 119.16 g/mol

Tetramethylammonium formate may provide the lowest temperature sensitivity, additionally reducing corrosion of stainless-steel flow system by chloride-induced depassivation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59138-84-0
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00054402
InChI Key WWIYWFVQZQOECA-UHFFFAOYSA-M
Synonym tetramethylammonium formate, fibrogenina, forgenin, tonoformina, methanaminium, n,n,n-trimethyl-, formate, tetramethylazanium formate, n,n,n-trimethylmethanaminium formate, ammonium, tetramethyl-, formate, methanaminium, n,n,n-trimethyl-, formate 1:1, c4h12n.cho2
PubChem CID 42961
IUPAC Name tetramethylazanium;formate
SMILES [O-]C=O.C[N+](C)(C)C

Specifications

Linear Formula (CH3)4N(COOH)
Quantity 500 mL
Beilstein 3916177
Solubility Information Fully miscible in water.
Formula Weight 119.16
Concentration or Composition (by Analyte or Components) 30% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetramethylammonium formate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.