missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium chloride, 98+%

Product Code. 10031620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
250g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031620 250 g Plastic bottle 250g
10795971 100 g Plastic bottle 100g
10125493 1 kg Plastic bottle 1kg
13498348 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10031620 Supplier Thermo Scientific Acros Supplier No. 138332500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-57-0 | C4H12ClN | 109.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-57-0
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.6
MDL Number MFCD00011628
InChI Key OKIZCWYLBDKLSU-UHFFFAOYSA-M
Synonym tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
PubChem CID 6379
ChEBI CHEBI:7070
IUPAC Name tetramethylazanium;chloride
SMILES C[N+](C)(C)C.[Cl-]

Specifications

Melting Point 420°C
Color White to Ivory
Infrared Spectrum Authentic
Assay Percent Range 98% min. (Argentometry)
Packaging Plastic bottle
Linear Formula (CH3)4NCl
Quantity 250 g
Solubility Information Solubility in water: >60% (20°C). Other solubilities: insoluble in ether,benzene,slightly soluble in ethanol
Formula Weight 109.6
Vapor Pressure Negligible
Percent Purity 98+%
Physical Form Hygroscopic Crystalline Powder
Chemical Name or Material Tetramethylammonium chloride, 98+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethylammonium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H370-Causes damage to organs.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.