missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium Bromide, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Tetramethylammonium Bromide |
| CAS | 64-20-0 |
| Molecular Formula | C4H12BrN |
| Quantity | 250 mg |
| Synonym | Methanaminium','N,N,N-trimethyl-','bromide (1:1) (ACI), Ammonium','tetramethyl-','bromide (8CI), Methanaminium','N,N,N-trimethyl-','bromide (9CI), Methyl','(dimethylamino)-','methobromide (7CI), Tetramethylammonium bromide (6CI), TMAB |
| InChI Formula | InChI=1S/C4H12N.BrH/c1-5(2,3)4;/h1-4H3;1H/q+1;/p-1 |
| SMILES | [Br-].C[N+](C)(C)C |
| IUPAC Name | tetramethylazanium;bromide |
| Molecular Weight (g/mol) | 154.05 |
| Formula Weight | 153.0153 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction