missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium bromide, 98%

Product Code. 11451930
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451930 100 g 100g
11461930 500 g 500g
15403077 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451930 Supplier Thermo Scientific Alfa Aesar Supplier No. A13368.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64-20-0 | C4H12BrN | 154.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Tetramethylammonium bromide acts as an ion pair reagent. It is used as a surface-active agent and an active ingredient for conditioners. Further, it is used as a softner for textiles and paper products. It is also employed as a disinfection agent, sanitizer, antimicrobial and antistatic agent. In addition to this, it plays in important role as a phase transfer catalyst in synthetic chemistry.

Solubility
Soluble in water, alcohol, chloroform and acetone.

Notes
Hygroscopic. Incompatible with oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-20-0
Molecular Formula C4H12BrN
Molecular Weight (g/mol) 154.05
MDL Number MFCD00011626
InChI Key DDFYFBUWEBINLX-UHFFFAOYSA-M
Synonym tetramethylammonium bromide, tetramethylammoniumbromide, tetramethylazanium bromide, methanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylmethanaminium bromide, unii-6l9ne9609p, ammonium, tetramethyl-, bromide, methanaminium, n,n,n-trimethyl-, bromide 1:1, c4h12n.br, acmc-209nkm
PubChem CID 66137
ChEBI CHEBI:55317
IUPAC Name tetramethylazanium;bromide
SMILES [Br-].C[N+](C)(C)C

Specifications

Melting Point >300°C
Density 1.56
Odor Odorless
Linear Formula (CH3)4NBr
Quantity 100 g
UN Number UN2811
Beilstein 3620955
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol,chloroform and acetone.
Formula Weight 154.06
Percent Purity 98%
Chemical Name or Material Tetramethylammonium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.