missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium acetate, tech. 90%

Product Code. 11416640
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416640 25 g 25g
11406640 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416640 Supplier Thermo Scientific Alfa Aesar Supplier No. H27659.14
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 10581-12-1 | C6H15NO2 | 133.19 g/mol

Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylammonium acetate is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. Further, it is used to prepare (1R,4S)-4-hydroxycyclopent-2-en-1-essigsaeure-methylester by reacting with 1,3-dimethyl-imidazolidin-2-one. In addition to this, it serves as an emulsifier, catalyst, sterilizer, antistatic and antiseptic agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10581-12-1
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00011630
InChI Key MRYQZMHVZZSQRT-UHFFFAOYSA-M
Synonym tetramethylammonium acetate, methanaminium, n,n,n-trimethyl-, acetate, tetramethyl ammonium acetate, methanaminium, n,n,n-trimethyl-, acetate 1:1, tetramethylammonium ion acetate, ammonium, tetramethyl-, acetate, tetramethylazanium acetate, n,n,n-trimethyl-methanaminiuacetate, acetate ion; tetramethylammonium ion
PubChem CID 82741
IUPAC Name tetramethylazanium acetate
SMILES CC([O-])=O.C[N+](C)(C)C

Specifications

Melting Point 256°C (decomposition)
Linear Formula (CH3)4N(OCOCH3)
Quantity 25 g
Beilstein 3571830
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 133.2
Percent Purity 90%
Grade Technical
Chemical Name or Material Tetramethylammonium acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.