missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyl orthosilicate, 99%

Product Code. 10308010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10308010 25 g Glass bottle 25g
10062620 100 g Glass bottle 100g
10154360 500 g Glass bottle 500g
11955161 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10308010 Supplier Thermo Scientific Supplier No. 203820250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 681-84-5 | C4H12O4Si | 152.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 681-84-5
Molecular Formula C4H12O4Si
Molecular Weight (g/mol) 152.22
MDL Number MFCD00008341
InChI Key LFQCEHFDDXELDD-UHFFFAOYSA-N
Synonym tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate
PubChem CID 12682
IUPAC Name tetramethyl silicate
SMILES CO[Si](OC)(OC)OC

Specifications

Melting Point -2.0°C
Density 1.0300g/mL
Boiling Point 121.0°C to 122.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Si(OCH3)4
Refractive Index 1.3670 to 1.3700
Quantity 25 g
Beilstein 01,287
Specific Gravity 1.03
Solubility Information Solubility in water: hydrolysis in water. Other solubilities: soluble in alcohol
Viscosity 0.7 mPa.s (20°C)
Formula Weight 152.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetramethyl orthosilicate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethyl orthosilicate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.