missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyl orthocarbonate, 99%

Product Code. 15515046 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15515046 5 g 5g
15525046 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15515046 Supplier Thermo Scientific Acros Supplier No. 158110050
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1850-14-2 | C5H12O4 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1850-14-2
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.15
InChI Key AHJWSRRHTXRLAQ-UHFFFAOYSA-N
Synonym tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc
PubChem CID 74613
IUPAC Name tetramethoxymethane
SMILES COC(OC)(OC)OC

Specifications

Melting Point -5.0°C
Density 1.0200g/mL
Boiling Point 114.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula C(OCH3)4
Refractive Index 1.3845
Quantity 5 g
Beilstein 03,II,4
Fieser 03,285
Specific Gravity 1.02
Formula Weight 136.15
Percent Purity 99%
Chemical Name or Material Tetramethyl orthocarbonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetramethyl orthocarbonate, 99%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.