missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyl methylenediphosphonate, 98+%

Product Code. 11475493
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475493 100 g 100g
11455493 5 g 5g
11465493 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475493 Supplier Thermo Scientific Alfa Aesar Supplier No. A13441.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16001-93-7 | C5H14O6P2 | 232.109 g/mol

Tetramethyl methylenediphosphonate is used in the preparation of phosphonate analogue of ribose-1-phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl methylenediphosphonate is used in the preparation of phosphonate analogue of ribose-1-phosphate.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16001-93-7
Molecular Formula C5H14O6P2
Molecular Weight (g/mol) 232.109
MDL Number MFCD00014884
InChI Key XAVFZUKFLWOSOS-UHFFFAOYSA-N
Synonym tetramethyl methylenediphosphonate, bis dimethoxyphosphoryl methane, tetramethyl methylenebis phosphonate, phosphonic acid, methylenebis-, tetramethyl ester, acmc-20ajjd, tetramethyl methanebisphosphonate, tetramethyl-methylenediphosphonate, tetramethyl methylenebisphosphonate, methylenebis phosphonic acid dimethyl ester, methylenediphosphonic acid tetramethyl ester
PubChem CID 519206
IUPAC Name bis(dimethoxyphosphoryl)methane
SMILES COP(=O)(CP(=O)(OC)OC)OC

Specifications

Boiling Point 152°C to 154°C (2.5 mmHg)
Refractive Index 1.455
Quantity 100 g
Beilstein 1874756
Solubility Information Miscible with water.
Formula Weight 232.11
Percent Purity ≥98%
Chemical Name or Material Tetramethyl methylenediphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.