missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyl Methylenebis(phosphonate), TRC

Product Code. 30452499
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452499 100 mg 100mg
30450589 50 mg 50mg
30450491 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30452499 Supplier Toronto Research Chemicals Supplier No. TRCT889680100MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16001-93-7
Molecular Formula C5H14O6P2
Molecular Weight (g/mol) 232.1
IUPAC Name bis(dimethoxyphosphoryl)methane
SMILES COP(=O)(CP(=O)(OC)OC)OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H14O6P2/c1-8-12(6,9-2)5-13(7,10-3)11-4/h5H2,1-4H3
Formula Weight 232.0266
Chemical Name or Material Tetramethyl Methylenebis(phosphonate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.