missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyl Ammonium Hydrogen Sulfate, for HPLC, Certified, Fisher Chemical

Product Code. 10090410 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Jar
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10090410 5 g Glass Jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10090410 Supplier Fisher Chemical Supplier No. T/0935/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 295°C
CAS 80526-82-5
Color White
Physical Form Powder/Solid
Quantity 5 g
Molecular Formula C4H13NO4S
MDL Number 36149
Synonym tetramethylammonium hydrogen sulfate, tetramethylammonium bisulfate, tetramethylammonium hydrogen sulphate, tetramethylammonium hydrogensulfate, hydrogen sulfate; tetramethylazanium, methanaminium, n,n,n-trimethyl-, sulfate 1:1, hydrogen sulfate; tetramethylammonium, acmc-209pjn, ch3 4n hso4 .h2o, tetramethylammonium bisulfate solution
InChI Key DWTYPCUOWWOADE-UHFFFAOYSA-M
SMILES C[N+](C)(C)C.OS(=O)(=O)[O-]
IUPAC Name hydrogen sulfate;tetramethylazanium
Molecular Weight (g/mol) 171.211
PubChem CID 157340
Formula Weight 171.21g/mol
Packaging Glass Jar
Show More Show Less
compliance-icons
Product Identifier
  • Tetramethylammonium hydrogensulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.