missing translation for 'onlineSavingsMsg'
Learn More

Tetramethoxysilane, 98%

Product Code. 15468909
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468909 500 g 500g
11456760 25 g 25g
11466760 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15468909 Supplier Thermo Scientific Alfa Aesar Supplier No. L09716.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 681-84-5 | C4H12O4Si | 152.22 g/mol

Tetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.

Solubility
Miscible with ethanol and organic solvents. Immiscible with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 681-84-5
Molecular Formula C4H12O4Si
Molecular Weight (g/mol) 152.22
MDL Number MFCD00008341
InChI Key LFQCEHFDDXELDD-UHFFFAOYSA-N
Synonym tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate
PubChem CID 12682
IUPAC Name tetramethyl silicate
SMILES CO[Si](OC)(OC)OC

Specifications

Boiling Point 121°C to 122°C
Melting Point 4°C to 5°C
Quantity 500 g
UN Number UN2606
Beilstein 1699658
Flash Point 26°C (78°F)
Solubility Information Miscible with ethanol and organic solvents. Immiscible with water.
Formula Weight 152.22
Percent Purity 98%
Odor Ether-like
Refractive Index 1.368
Sensitivity Moisture sensitive
Density 1.032
Chemical Name or Material Tetramethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.