missing translation for 'onlineSavingsMsg'
Learn More

Tetramethoxymethane, TRC

Product Code. 30412769
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412769 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412769 Supplier Toronto Research Chemicals Supplier No. TRCT2988605G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetramethoxymethane
CAS 1850-14-2
Molecular Formula C5H12O4
Quantity 5 g
Synonym Tetramethyl Ester Orthocarbonic Acid, Dimethoxymethylal, NSC 359558, Tetramethoxymethane, Tetramethyl orthocarbonate
InChI Formula InChI=1S/C5H12O4/c1-6-5(7-2,8-3)9-4/h1-4H3
SMILES COC(OC)(OC)OC
IUPAC Name tetramethoxymethane
Molecular Weight (g/mol) 136.15
Formula Weight 136.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.