missing translation for 'onlineSavingsMsg'
Learn More

Tetrakisepoxy cyclosiloxane

Product Code. 15492807
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492807 25 g 25g
15402817 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492807 Supplier Thermo Scientific Alfa Aesar Supplier No. 046960.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 257284-60-9 | C28H56O12Si4 | 697.084 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 257284-60-9
Molecular Formula C28H56O12Si4
Molecular Weight (g/mol) 697.084
MDL Number MFCD22380706
InChI Key BLZDTVROPKIJLZ-UHFFFAOYSA-N
Synonym 2,4,6,8-tetramethyl-2,4,6,8-tetrakis 3-oxiranylmethoxy propyl cyclotetrasiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetrakis 3-oxiran-2-ylmethoxy propyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, tetrakisepoxy cyclosiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetrakis 3-propyl cyclotetrasiloxane, 2,4,6,8-tetrakis 6,7-epoxy-4-oxaheptyl-2,4,6,8-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetrakis propyl glycidyl ether cyclotetrasiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetrakis 3-glycidyloxy propyl cyclooctanetetrasiloxane, cyclotetrasiloxane,2,4,6,8-tetramethyl-2,4,6,8-tetrakis 3-2-oxiranylmethoxy propyl
PubChem CID 108490
IUPAC Name 2,4,6,8-tetramethyl-2,4,6,8-tetrakis[3-(oxiran-2-ylmethoxy)propyl]-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
SMILES C[Si]1(O[Si](O[Si](O[Si](O1)(C)CCCOCC2CO2)(C)CCCOCC3CO3)(C)CCCOCC4CO4)CCCOCC5CO5

Specifications

Color Colorless
Physical Form Colorless liquid
Quantity 25 g
Formula Weight 697.09
Chemical Name or Material Tetrakisepoxy cyclosiloxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.