missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(triphenylphosphine)platinum(0), Pt 15.2% min

Product Code. 11396058
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396058 5 g 5g
11306068 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396058 Supplier Thermo Scientific Alfa Aesar Supplier No. 010549.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrosilation and oxidation | CAS: 14221-02-4 | C72H60P4Pt | 1244.25 g/mol

Tetrakis(triphenylphosphine)platinum(0), Pt is widely used in hydrosilation and oxidation. In Catalysts, Metal & Specialty Catalysts, Products & Industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 14221-02-4
Molecular Formula C72H60P4Pt
Molecular Weight (g/mol) 1244.25
MDL Number MFCD00010014
InChI Key SYKXNRFLNZUGAJ-UHFFFAOYSA-N
Synonym tetrakis triphenylphosphine platinum 0, tetrakis triphenylphosphine platinum, platinum; triphenylphosphane, pt pph3 4, platinum tetratriphenylphosphine, platinum, tetrakis triphenylphosphine-, t-4, platinum 0-tetrakis triphenylphosphine, platinum; triphenylphosphine, tetrakis triphenylphosphine platinum 0 1g
PubChem CID 11979705
IUPAC Name platinum;triphenylphosphane
SMILES [Pt].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Spécification

Color Yellow
Odor Odorless
Quantity 5 g
Sensitivity Air sensitive
Solubility Information Soluble in benzene
Formula Weight 1244.21
Concentration or Composition (by Analyte or Components) Pt 15.2% min.
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(triphenylphosphine)platinum(0)

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.