missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(trimethylsilyl)silane, 98%

Product Code. 11490097
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11490097

Brand: Thermo Scientific Alfa Aesar B21020.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4098-98-0 | C12H36Si5 | 320.85 g/mol

It is used as a starting material for the synthesis of tris(trimethylsilyl)silyl lithium with methyl lithium and in the preparation of lithium bis[(TMS)3silyl]cuprate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a starting material for the synthesis of tris(trimethylsilyl)silyl lithium with methyl lithium and in the preparation of lithium bis[(TMS)3silyl]cuprate.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4098-98-0
Molecular Formula C12H36Si5
Molecular Weight (g/mol) 320.85
MDL Number MFCD00054859
InChI Key BOJSDHZZKKYWAS-UHFFFAOYSA-N
Synonym tetrakis trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl trisilane, tetrakil trimethylsilyl silane, si si ch3 3 4, trisilane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl, 2,2-bis trimethylsilyl-1,1,1,3,3,3-hexamethyl-trisilane, acmc-1an6g, tetra trimethylsilyl-silane, bojsdhzzkkywas-uhfffaoysa
PubChem CID 138115
IUPAC Name tetrakis(trimethylsilyl)silane
SMILES C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C

Specifications

Melting Point 266°C to 268°C (sublimation)
Quantity 1 g
Formula Weight 320.85
Percent Purity 98%
Chemical Name or Material Tetrakis(trimethylsilyl)silane
compliance-icons
Product Identifier
  • Tetrakis(trimethylsilyl)silane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Tetrakis(trimethylsilyl)silane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.