missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylsiloxy)silane, 97%

Product Code. 11490188
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490188 5 g 5g
11400198 25 g 25g
11410198 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490188 Supplier Thermo Scientific Alfa Aesar Supplier No. B23754.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17082-47-2 | C8H24O4Si5 | 324.70 g/mol

It is employed for non-volatile NMR standard for high temperature work, in place of the highly volatile and as cross linker for vinyl functional silicones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed for non-volatile NMR standard for high temperature work, in place of the highly volatile and as cross linker for vinyl functional silicones.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17082-47-2
Molecular Formula C8H24O4Si5
Molecular Weight (g/mol) 324.70
MDL Number MFCD00053848
InChI Key UOUILILVWRHZSH-UHFFFAOYSA-N
Synonym tetrakis dimethylsilyl orthosilicate

Specifications

Boiling Point 188°C to 190°C
Melting Point -60°C
Quantity 5 g
Flash Point 67°C (153°F)
Refractive Index 1.388
Sensitivity Moisture sensitive
Density 0.884
Beilstein 1793898
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 328.73
Percent Purity 97%
Chemical Name or Material Tetrakis(dimethylsiloxy)silane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.