missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)titanium(IV), 99.9% (metals basis)

Product Code. 11360198
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11360198 5 g 5g
11380198 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11360198 Supplier Thermo Scientific Alfa Aesar Supplier No. 042932.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3275-24-9 | C8H24N4Ti | 224.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3275-24-9
Molecular Formula C8H24N4Ti
Molecular Weight (g/mol) 224.18
MDL Number MFCD00014861
InChI Key MNWRORMXBIWXCI-UHFFFAOYSA-N
Synonym tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva
PubChem CID 123185
IUPAC Name dimethylazanide;titanium(4+)
SMILES [Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C

Specifications

Color Brown-Yellow
Density 0.96 g/mL
Boiling Point 50°C (0.5 mmHg)
Flash Point −30°C (−22°F)
Linear Formula Ti[N(CH3)2]4
Quantity 5 g
UN Number UN3129
Sensitivity Moisture sensitive
Formula Weight 224.19
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tetrakis (dimethylamino)titanium (IV)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.