missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)germanium(IV), 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11355567
Change view
Click to view available options
Quantity:
500 mg
2 g
10 g
Unit Size:
10g
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355567 500 mg 500mg
11365567 2 g 2g
11345567 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11355567 Supplier Thermo Scientific Alfa Aesar Supplier No. 042760.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Preparation of nitrides

Preparation of nitrides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7344-40-3
Molecular Formula C8H24GeN4
Molecular Weight (g/mol) 248.938
MDL Number MFCD01863476
InChI Key JKUUTODNPMRHHZ-UHFFFAOYSA-N
Synonym germane, tetrakis dimethylamino, octamethylgermanetetramine, tetrakis dimethylamino germane, tetrakis dimethylamino germanium, dimethyl tris dimethylamino germyl amine
PubChem CID 16684231
IUPAC Name N-methyl-N-[tris(dimethylamino)germyl]methanamine
SMILES CN(C)[Ge](N(C)C)(N(C)C)N(C)C

Specifications

Melting Point 14°C
Color Colorless
Density 1.07 g/mL
Boiling Point 203°C
Flash Point 34°C (93°F)
Odor Amine-like
Linear Formula Ge[N(CH3)2]4
Quantity 500 mg
UN Number UN3129
Sensitivity Moisture sensitive
Formula Weight 248.9
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tetrakis (dimethylamino)germanium (IV)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.